missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,5-Dinitrobenzoyl chloride, 98+%
Description
Used in photography. This aromatic compound is used by chemists to identify alcohol components in esters and in the fluorometric analysis of creatinine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Used in photography. This aromatic compound is used by chemists to identify alcohol components in esters and in the fluorometric analysis of creatinine.
Solubility
Slightly soluble in water.
Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, bases, water.
Used in photography. This aromatic compound is used by chemists to identify alcohol components in esters and in the fluorometric analysis of creatinine.
Solubility
Slightly soluble in water.
Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, bases, water.
Specifications
Specifications
| Melting Point | 67°C to 70°C |
| Boiling Point | 196°C (12 mmHg) |
| Quantity | 25 g |
| UN Number | UN3261 |
| Beilstein | 990249 |
| Sensitivity | Moisture sensitive |
| Merck Index | 14,3277 |
| Solubility Information | Slightly soluble in water. |
| Formula Weight | 230.57 |
| Percent Purity | ≥98% |
| Show More |
Safety and Handling
| Hazard Category | H314-H341 |
| Hazard Statement | GHS H Statement H314-H318 Causes severe skin burns and eye damage. Causes serious eye damage. |
| Precautionary Statement | P201-P202-P260-P264b-P281-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
| DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S. |
| EINECSNumber | 202-750-6 |
| RTECSNumber | DM6637000 |
| TSCA | Yes |
| Recommended Storage | Keep cold; Store under Nitrogen |
RUO – Research Use Only