missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dinitrobenzoic Acid 98.0+%, TCI America™

Supplier:  TCI America D0824500G

 View more versions of this product

Catalog No. D0824500G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

3,5-Dinitrobenzoic Acid
207°C
C7H4N2O6
500 g
VYWYYJYRVSBHJQ-UHFFFAOYSA-N
3,5-dinitrobenzoic acid
7433
212.12
Crystalline Powder
99-34-3
White-Yellow
MFCD00007253
3,5-dinitrobenzoic acid, dinitrobenzoic acid, dnba, benzoic acid, 3,5-dinitro, 3-carboxy-1,5-dinitrobenzene, 3,5-dinitro-benzoic acid, unii-4v3f9q018p, ccris 3129, 3,5-dnba, 3,5-dinitrobenzenecarboxylic acid
OC(=O)C1=CC(=CC(=C1)[N+]([O-])=O)[N+]([O-])=O
212.12
CHEBI:73914
≥98.0% (GC,T)

Chemical Identifiers

99-34-3
212.12
VYWYYJYRVSBHJQ-UHFFFAOYSA-N
7433
3,5-dinitrobenzoic acid
C7H4N2O6
MFCD00007253
3,5-dinitrobenzoic acid, dinitrobenzoic acid, dnba, benzoic acid, 3,5-dinitro, 3-carboxy-1,5-dinitrobenzene, 3,5-dinitro-benzoic acid, unii-4v3f9q018p, ccris 3129, 3,5-dnba, 3,5-dinitrobenzenecarboxylic acid
CHEBI:73914
OC(=O)C1=CC(=CC(=C1)[N+]([O-])=O)[N+]([O-])=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-1505&(3)-1511

RTECSNumber : DG9140700

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions