missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dinitrobenzoic Acid 98.0+%, TCI America™

Catalog No. D082425G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D082425G 25 g
D0824500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D082425G Supplier TCI America Supplier No. D082425G

Chemical Identifiers

CAS 99-34-3
Molecular Formula C7H4N2O6
Molecular Weight (g/mol) 212.12
MDL Number MFCD00007253
InChI Key VYWYYJYRVSBHJQ-UHFFFAOYSA-N
Synonym 3,5-dinitrobenzoic acid, dinitrobenzoic acid, dnba, benzoic acid, 3,5-dinitro, 3-carboxy-1,5-dinitrobenzene, 3,5-dinitro-benzoic acid, unii-4v3f9q018p, ccris 3129, 3,5-dnba, 3,5-dinitrobenzenecarboxylic acid
PubChem CID 7433
ChEBI CHEBI:73914
IUPAC Name 3,5-dinitrobenzoic acid
SMILES OC(=O)C1=CC(=CC(=C1)[N+]([O-])=O)[N+]([O-])=O

Specifications

Melting Point 207°C
Color White-Yellow
Quantity 25 g
Formula Weight 212.12
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dinitrobenzoic Acid
EINECSNumber (3)-1505&(3)-1511
RTECSNumber DG9140700
TSCA Yes