Learn More
3,5-Dinitrobenzoic acid, 98+%
CAS: 99-34-3 | C7H4N2O6 | 212.12 g/mol
Supplier: Thermo Scientific Chemicals A165020B
Description
3,5-Dinitrobenzoic acid, is used as a reagent used in the derivatization of resins and the determination of ampicillin.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3,5-Dinitrobenzoic acid, is used as a reagent used in the derivatization of resins and the determination of ampicillin.
Solubility
It is Soluble in ethanol (50 mg/mL), methanol, alcohol (very), and glacial acetic acid (very). Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agents, Strong bases, Strong reducing agents.
Specifications
| 3,5-Dinitrobenzoic acid | |
| 204°C to 208°C | |
| C7H4N2O6 | |
| 1000 g | |
| 1914286 | |
| 3,5-dinitrobenzoic acid, dinitrobenzoic acid, dnba, benzoic acid, 3,5-dinitro, 3-carboxy-1,5-dinitrobenzene, 3,5-dinitro-benzoic acid, unii-4v3f9q018p, ccris 3129, 3,5-dnba, 3,5-dinitrobenzenecarboxylic acid | |
| VYWYYJYRVSBHJQ-UHFFFAOYSA-N | |
| 3,5-dinitrobenzoic acid | |
| 7433 | |
| 212.12 |
| 99-34-3 | |
| 1.68 | |
| MFCD00007253 | |
| UN1325 | |
| 14,3276 | |
| It is Soluble in ethanol (50mg/mL),methanol,alcohol (very),and glacial acetic acid (very). Insoluble in water. | |
| OC(=O)C1=CC(=CC(=C1)[N+]([O-])=O)[N+]([O-])=O | |
| 212.12 | |
| CHEBI:73914 | |
| ≥98% |
Chemical Identifiers
| 99-34-3 | |
| 212.12 | |
| VYWYYJYRVSBHJQ-UHFFFAOYSA-N | |
| 7433 | |
| 3,5-dinitrobenzoic acid |
| C7H4N2O6 | |
| MFCD00007253 | |
| 3,5-dinitrobenzoic acid, dinitrobenzoic acid, dnba, benzoic acid, 3,5-dinitro, 3-carboxy-1,5-dinitrobenzene, 3,5-dinitro-benzoic acid, unii-4v3f9q018p, ccris 3129, 3,5-dnba, 3,5-dinitrobenzenecarboxylic acid | |
| CHEBI:73914 | |
| OC(=O)C1=CC(=CC(=C1)[N+]([O-])=O)[N+]([O-])=O |
Safety and Handling
GHS H Statement
H301-H228-H315-H319-H335
Toxic if swallowed.
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
EINECSNumber : 202-751-1
RTECSNumber : DG9140700
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only