missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dinitroaniline, 98%

Catalog No. p-3211822
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC116940010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC116940010 Supplier Thermo Scientific Chemicals Supplier No. 116940010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 618-87-1 | C6H5N3O4 | 183.123 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 618-87-1
MDL Number MFCD00007263
InChI Key MPBZUKLDHPOCLS-UHFFFAOYSA-N
Synonym benzenamine, 3,5-dinitro, aniline, 3,5-dinitro, unii-0ahr6k1n73, ccris 3109, 3,5-dinitrophenylamine, 3-5-dinitro-phenylamine, 3,5-dinitroanilin, aniline,5-dinitro, acmc-209mx8, benzenamine,3,5-dinitro
PubChem CID 12068
IUPAC Name 3,5-dinitroaniline
SMILES C1=C(C=C(C=C1[N+](=O)[O-])[N+](=O)[O-])N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 159°C to 163°C
Color Brown-Yellow to Yellow
Flash Point >100°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Molecular Formula C6H5N3O4
Linear Formula (O2N)2C6H3NH2
Quantity 1 g
Beilstein 12, 759
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in organic solvents,soluble in ethanol and diethylether
Molecular Weight (g/mol) 183.12
Formula Weight 183.12
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 3, 5-Dinitroaniline
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Toxic in contact with skin.
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 210-567-8

RUO – Research Use Only