missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethylthiophenol, 97%

Catalog No. p-7044830
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0840306 5 g
AAL0840314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0840306 Supplier Thermo Scientific Chemicals Supplier No. L0840306

CAS: 38360-81-5 | C8H10S | 138.228 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 38360-81-5
Molecular Formula C8H10S
Molecular Weight (g/mol) 138.228
MDL Number MFCD00040226
InChI Key CESBAYSBPMVAEI-UHFFFAOYSA-N
Synonym 3,5-dimethylthiophenol, benzenethiol, 3,5-dimethyl, 3,5-dimethylbenzene-1-thiol, 3,5-dimethyl thiophenol, m-xylene-5-thiol, 5-mercapto-m-xylene, 3,5-xylenethiol, xylenethiol, mixed isomers, 3,5dimethylthiophenol, pubchem6885
PubChem CID 117548
IUPAC Name 3,5-dimethylbenzenethiol
SMILES CC1=CC(=CC(=C1)S)C

Specifications

Density 1.015
Boiling Point 127°C (50 mmHg)
Flash Point 85°C (185°F)
Refractive Index 1.5675
Quantity 5 g
UN Number UN2810
Beilstein 3233085
Sensitivity Air Sensitive
Formula Weight 138.23
Percent Purity 97%
Chemical Name or Material 3,5-Dimethylthiophenol
Show More Show Less
Hazard Category H227-H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H227-H302-H312-H315-H319-H335
Toxic if inhaled.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 253-901-8
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only