missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,5-Dimethylpyridine N-Oxide 98.0+%, TCI America™
Supplier: TCI America D546925G
Specifications
| 3,5-Dimethylpyridine N-Oxide | |
| 106°C | |
| 168°C | |
| MFCD00126839 | |
| 3,5-Lutidine N-Oxide | |
| CC1=CC(=C[N+](=C1)[O-])C | |
| 123.155 | |
| 123.16 | |
| Crystalline Powder |
| 3718-65-8 | |
| White-Yellow | |
| C7H9NO | |
| 25 g | |
| SSTLCMOZTCLOLQ-UHFFFAOYSA-N | |
| 3,5-dimethyl-1-oxidopyridin-1-ium | |
| 19485 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 3718-65-8 | |
| 123.155 | |
| SSTLCMOZTCLOLQ-UHFFFAOYSA-N | |
| 19485 | |
| CC1=CC(=C[N+](=C1)[O-])C |
| C7H9NO | |
| MFCD00126839 | |
| 3,5-Lutidine N-Oxide | |
| 3,5-dimethyl-1-oxidopyridin-1-ium |
Safety and Handling
TSCA : No