missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethylpyridine N-Oxide 98.0+%, TCI America™

Catalog No. p-7128781
Change view
Click to view available options
Quantity:
25 g
5 g
Catalog No. Quantity
D54695G 5 g
D546925G 25 g
2 options

Catalog No. D54695G

Supplier: TCI America D54695G

Only null left

Chemical Identifiers

CAS 3718-65-8
Molecular Formula C7H9NO
Molecular Weight (g/mol) 123.155
MDL Number MFCD00126839
InChI Key SSTLCMOZTCLOLQ-UHFFFAOYSA-N
Synonym 3,5-Lutidine N-Oxide
PubChem CID 19485
IUPAC Name 3,5-dimethyl-1-oxidopyridin-1-ium
SMILES CC1=CC(=C[N+](=C1)[O-])C

Specifications

Melting Point 106°C
Color White-Yellow
Boiling Point 168°C
Quantity 5 g
Formula Weight 123.16
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dimethylpyridine N-Oxide