missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethylphenylacetic acid, 98+%

Catalog No. AAL0970703
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0970703 1 g
AAL0970706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0970703 Supplier Thermo Scientific Chemicals Supplier No. L0970703
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 42288-46-0 | C10H11O2 | 163.20 g/mol

It is an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an active pharmaceutical intermediate.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 42288-46-0
Molecular Formula C10H11O2
Molecular Weight (g/mol) 163.20
MDL Number MFCD00082776
InChI Key HDNBKTWQBJJYPD-UHFFFAOYSA-M
Synonym 3,5-dimethylphenylacetic acid, 2-3,5-dimethylphenyl acetic acid, 3,5-dimethylphenyl acetic acid, benzeneacetic acid, 3,5-dimethyl, 3,5-xylyl acetic acid, acmc-20anc9, ksc238q4h, 3,5-dimethylbenzeneacetic acid, 3,5-dimethyl-phenyl-acetic acid
PubChem CID 4749495
IUPAC Name 2-(3,5-dimethylphenyl)acetic acid
SMILES CC1=CC(CC([O-])=O)=CC(C)=C1

Specifications

Melting Point 99°C to 103°C
Quantity 1 g
Beilstein 2083903
Formula Weight 164.21
Percent Purity ≥98%
Chemical Name or Material 3,5-Dimethylphenylacetic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only