missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethylphenyl Isocyanate 98.0+%, TCI America™

Catalog No. p-7127748
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D34745G 5 g
D347425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D34745G Supplier TCI America Supplier No. D34745G
Only null left

Chemical Identifiers

CAS 54132-75-1
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.177
MDL Number MFCD00013868
InChI Key DZSGDHNHQAJZCO-UHFFFAOYSA-N
Synonym 3,5-dimethylphenyl isocyanate, 3,5-dimethylphenylisocyanate, 3,5-xylyl isocyanate, benzene, 1-isocyanato-3,5-dimethyl, isocyanic acid 3,5-dimethylphenyl ester, 5-isocyanato-m-xylene, 1-isocyanato-3,5-dimethyl-benzene, acmc-1axct, 3,5-dimethylbenzenisocyanate, ksc274i2t
PubChem CID 521488
IUPAC Name 1-isocyanato-3,5-dimethylbenzene
SMILES CC1=CC(=CC(=C1)N=C=O)C

Specifications

Color Yellow
Boiling Point 205°C
Quantity 5 g
Physical Form Clear Liquid at 20°C
UN Number 2206
Formula Weight 147.18
Percent Purity ≥98.0% (GC)
Chemical Name or Material 3,5-Dimethylphenyl Isocyanate
TSCA No
Recommended Storage Refrigerator