missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethylphenyl isocyanate, 97%

Catalog No. AAL1169806
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1169806 5 g
AAL1169803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1169806 Supplier Thermo Scientific Chemicals Supplier No. L1169806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 54132-75-1 | C9H9NO | 147.177 g/mol

3,5-Dimethylphenyl isocyanate is applications include linking with oligosaccharides and cyclodextrins for chiral chromatography stationary phases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Dimethylphenyl isocyanate is applications include linking with oligosaccharides and cyclodextrins for chiral chromatography stationary phases.

Solubility
Hydrolyzes in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 54132-75-1
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.177
MDL Number MFCD00013868
InChI Key DZSGDHNHQAJZCO-UHFFFAOYSA-N
Synonym 3,5-dimethylphenyl isocyanate, 3,5-dimethylphenylisocyanate, 3,5-xylyl isocyanate, benzene, 1-isocyanato-3,5-dimethyl, isocyanic acid 3,5-dimethylphenyl ester, 5-isocyanato-m-xylene, 1-isocyanato-3,5-dimethyl-benzene, acmc-1axct, 3,5-dimethylbenzenisocyanate, ksc274i2t
PubChem CID 521488
IUPAC Name 1-isocyanato-3,5-dimethylbenzene
SMILES CC1=CC(=CC(=C1)N=C=O)C

Specifications

Density 1.045
Quantity 5 g
Refractive Index 1.528
UN Number UN2206
Beilstein 774958
Boiling Point 205°C
Solubility Information Hydrolyzes in water.
Formula Weight 147.18
Percent Purity 97%
Chemical Name or Material 3,5-Dimethylphenyl isocyanate
Hazard Category H227-H302+H312+H332-H315-H319-H334-H335
Hazard Statement GHS H Statement
H300-H330-H334-H227-H315-H319-H335
Fatal if swallowed.
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P342+P311-P362-P370+P378q-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 258-987-0
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only