missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethyloctane 98.0+%, TCI America™

Catalog No. D12145ML
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D12145ML 5 mL
D12141ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D12145ML Supplier TCI America Supplier No. D12145ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 15869-93-9
Molecular Formula C10H22
Molecular Weight (g/mol) 142.29
MDL Number MFCD00048739
InChI Key VRHRGVJOUHJULC-UHFFFAOYNA-N
PubChem CID 139989
ChEBI CHEBI:84237
IUPAC Name 3,5-dimethyloctane
SMILES CCCC(C)CC(C)CC

Specifications

Color Colorless
Boiling Point 160°C
Quantity 5 mL
UN Number 3295
Formula Weight 142.29
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,5-Dimethyloctane
EINECSNumber (2)-0010
TSCA No