accessibility menu, dialog, popup

UserName

CAS RN 15869-93-9

IUPAC Name: 3,5-dimethyloctane
Synonyms: octane, 3,5-dimethyl-
Molecular Weight (g/mol): 142.29
Molecular Formula: C10H22
InChi Key: VRHRGVJOUHJULC-UHFFFAOYNA-N
SMILES: CCCC(C)CC(C)CC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results