missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethylisoxazole-4-carboxaldehyde, 97%

Catalog No. aah6191214
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6191214 25 g
AAH6191206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6191214 Supplier Thermo Scientific Chemicals Supplier No. H6191214
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 54593-26-9 | C6H7NO2 | 125.127 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 54593-26-9
Molecular Formula C6H7NO2
Molecular Weight (g/mol) 125.127
MDL Number MFCD02681977
InChI Key TVAYXKLCEILMEA-UHFFFAOYSA-N
Synonym 3,5-dimethyl-4-isoxazolecarbaldehyde, 3,5-dimethylisoxazole-4-carbaldehyde, 3,5-dimethylisoxazole-4-carboxaldehyde, 4-isoxazolecarboxaldehyde, 3,5-dimethyl, 3,5-dimethyl-4-isoxazolecarboxaldehyde, 3,5-dimethyl-4-formalisoxazole, pubchem8682, acmc-1axd1, ksc496e3j, 3,5-dimethylisoxazol-4-carbaldehyde
PubChem CID 289576
IUPAC Name 3,5-dimethyl-1,2-oxazole-4-carbaldehyde
SMILES CC1=C(C(=NO1)C)C=O

Specifications

Melting Point 35°C to 38°C
Quantity 25 g
Formula Weight 125.13
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 3,5-Dimethylisoxazole-4-carboxaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only