missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethylbenzyl bromide, 98%

Catalog No. AAA1426706
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1426706 5 g
AAA1426714 25 g
AAA1426722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1426706 Supplier Thermo Scientific Chemicals Supplier No. A1426706

CAS: 27129-86-8 | C9H11Br | 199.091 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 27129-86-8
Molecular Formula C9H11Br
Molecular Weight (g/mol) 199.091
MDL Number MFCD00013539
InChI Key QXDHXCVJGBTQMK-UHFFFAOYSA-N
Synonym 3,5-dimethylbenzyl bromide, 1-bromomethyl-3,5-dimethylbenzene, 3,5-dimethylbenzylbromide, alpha-bromomesitylene, benzene, 1-bromomethyl-3,5-dimethyl, .alpha.-bromomesitylene, 5-bromomethyl-1,3-dimethylbenzene, a-bromomesitylene, pubchem9979, 3,5dimethylbenzyl bromide
PubChem CID 141334
IUPAC Name 1-(bromomethyl)-3,5-dimethylbenzene
SMILES CC1=CC(=CC(=C1)CBr)C

Specifications

Melting Point 36°C to 40°C
Boiling Point 109°C to 110°C (14 mmHg)
Quantity 5 g
UN Number UN3261
Beilstein 1857343
Formula Weight 199.1
Percent Purity 98%
Chemical Name or Material 3,5-Dimethylbenzyl bromide
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only