missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethylbenzoic Acid 99.0+%, TCI America™

Catalog No. p-7126371
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D068425G 25 g
D0684250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D068425G Supplier TCI America Supplier No. D068425G
Only null left

Chemical Identifiers

CAS 499-06-9
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.177
MDL Number MFCD00002525
InChI Key UMVOQQDNEYOJOK-UHFFFAOYSA-N
Synonym mesitylenic acid, benzoic acid, 3,5-dimethyl, unii-ed8av34n0y, 3,5-dimethyl benzoic acid, 3,5-dimethyl-benzoic acid, ed8av34n0y, 3,5-dimethylbenzoicacid, pubchem15440, 3,5-dimethylbezoic acid, 3,5 dimethylbenzoic acid
PubChem CID 10356
ChEBI CHEBI:64821
IUPAC Name 3,5-dimethylbenzoic acid
SMILES CC1=CC(=CC(=C1)C(=O)O)C

Specifications

Melting Point 170°C
Color White-Yellow
Quantity 25 g
Physical Form Crystal-Powder at 20°C
Formula Weight 150.18
Percent Purity ≥99.0% (T)
Chemical Name or Material 3,5-Dimethylbenzoic Acid
EINECSNumber (3)-4260
RTECSNumber DG8734030
TSCA Yes