Learn More
3,5-Dimethylbenzoic acid, 98+%
CAS: 499-06-9 | C9H10O2 | 150.177 g/mol
$70.26 - $1041.53
Chemical Identifiers
| CAS | 499-06-9 |
|---|---|
| Molecular Formula | C9H10O2 |
| Molecular Weight (g/mol) | 150.177 |
| MDL Number | MFCD00002525 |
| InChI Key | UMVOQQDNEYOJOK-UHFFFAOYSA-N |
| Synonym | mesitylenic acid, benzoic acid, 3,5-dimethyl, unii-ed8av34n0y, 3,5-dimethyl benzoic acid, 3,5-dimethyl-benzoic acid, ed8av34n0y, 3,5-dimethylbenzoicacid, pubchem15440, 3,5-dimethylbezoic acid, 3,5 dimethylbenzoic acid |
| PubChem CID | 10356 |
| ChEBI | CHEBI:64821 |
| IUPAC Name | 3,5-dimethylbenzoic acid |
| SMILES | CC1=CC(=CC(=C1)C(=O)O)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1119809
|
Thermo Scientific Chemicals
A1119809 |
10 g |
Each for $70.26
|
|
|||||
|
AAA1119818
|
Thermo Scientific Chemicals
A1119818 |
50 g |
Each for $248.90
|
|
|||||
|
AAA1119830
|
Thermo Scientific Chemicals
A1119830 |
250 g |
Each for $1,041.53
|
|
|||||
Description
3,5-Dimethylbenzoic acid is suitable reagent used as carbon and energy supplement in the culture medium of Pseudomonas sp
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3,5-Dimethylbenzoic acid is suitable reagent used as carbon and energy supplement in the culture medium of Pseudomonas sp
Solubility
Soluble in methanol. (1 g/10 mL). Slightly soluble in water.
Notes
Store away from oxidizing agents. Use only in a chemical fume hood. Incompatible with strong acids and bases.
Chemical Identifiers
| 499-06-9 | |
| 150.177 | |
| UMVOQQDNEYOJOK-UHFFFAOYSA-N | |
| 10356 | |
| 3,5-dimethylbenzoic acid |
| C9H10O2 | |
| MFCD00002525 | |
| mesitylenic acid, benzoic acid, 3,5-dimethyl, unii-ed8av34n0y, 3,5-dimethyl benzoic acid, 3,5-dimethyl-benzoic acid, ed8av34n0y, 3,5-dimethylbenzoicacid, pubchem15440, 3,5-dimethylbezoic acid, 3,5 dimethylbenzoic acid | |
| CHEBI:64821 | |
| CC1=CC(=CC(=C1)C(=O)O)C |
Specifications
| 168°C to 172°C | |
| 10 g | |
| C9H10O2 | |
| 1072182 | |
| Soluble in methanol. (1g/10 mL). Slightly soluble in water. | |
| CC1=CC(=CC(=C1)C(=O)O)C | |
| 150.177 | |
| CHEBI:64821 | |
| ≥98% |
| 499-06-9 | |
| Odorless | |
| MFCD00002525 | |
| mesitylenic acid, benzoic acid, 3,5-dimethyl, unii-ed8av34n0y, 3,5-dimethyl benzoic acid, 3,5-dimethyl-benzoic acid, ed8av34n0y, 3,5-dimethylbenzoicacid, pubchem15440, 3,5-dimethylbezoic acid, 3,5 dimethylbenzoic acid | |
| UMVOQQDNEYOJOK-UHFFFAOYSA-N | |
| 3,5-dimethylbenzoic acid | |
| 10356 | |
| 150.18 | |
| 3,5-Dimethylbenzoic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 207-876-5
RTECSNumber : DG8734030
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only