missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethylbenzoic acid, 98+%

Catalog No. AAA1119809
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1119809 10 g
AAA1119818 50 g
AAA1119830 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1119809 Supplier Thermo Scientific Chemicals Supplier No. A1119809
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 499-06-9 | C9H10O2 | 150.177 g/mol

3,5-Dimethylbenzoic acid is suitable reagent used as carbon and energy supplement in the culture medium of Pseudomonas sp

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Dimethylbenzoic acid is suitable reagent used as carbon and energy supplement in the culture medium of Pseudomonas sp

Solubility
Soluble in methanol. (1 g/10 mL). Slightly soluble in water.

Notes
Store away from oxidizing agents. Use only in a chemical fume hood. Incompatible with strong acids and bases.

Chemical Identifiers

CAS 499-06-9
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.177
MDL Number MFCD00002525
InChI Key UMVOQQDNEYOJOK-UHFFFAOYSA-N
Synonym mesitylenic acid, benzoic acid, 3,5-dimethyl, unii-ed8av34n0y, 3,5-dimethyl benzoic acid, 3,5-dimethyl-benzoic acid, ed8av34n0y, 3,5-dimethylbenzoicacid, pubchem15440, 3,5-dimethylbezoic acid, 3,5 dimethylbenzoic acid
PubChem CID 10356
ChEBI CHEBI:64821
IUPAC Name 3,5-dimethylbenzoic acid
SMILES CC1=CC(=CC(=C1)C(=O)O)C

Specifications

Melting Point 168°C to 172°C
Quantity 10 g
Odor Odorless
Beilstein 1072182
Solubility Information Soluble in methanol. (1g/10 mL). Slightly soluble in water.
Formula Weight 150.18
Percent Purity ≥98%
Chemical Name or Material 3,5-Dimethylbenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 207-876-5
RTECSNumber DG8734030
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only