missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethylbenzenethiol 97.0+%, TCI America™

Catalog No. D42865G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D42865G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D42865G Supplier TCI America Supplier No. D42865G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 38360-81-5
Molecular Formula C8H10S
Molecular Weight (g/mol) 138.228
MDL Number MFCD00040226
InChI Key CESBAYSBPMVAEI-UHFFFAOYSA-N
Synonym 3,5-dimethylthiophenol, benzenethiol, 3,5-dimethyl, 3,5-dimethylbenzene-1-thiol, 3,5-dimethyl thiophenol, m-xylene-5-thiol, 5-mercapto-m-xylene, 3,5-xylenethiol, xylenethiol, mixed isomers, 3,5dimethylthiophenol, pubchem6885
PubChem CID 117548
IUPAC Name 3,5-dimethylbenzenethiol
SMILES CC1=CC(=CC(=C1)S)C

Specifications

Color Yellow
Boiling Point 128°C
Quantity 5 g
Formula Weight 138.23
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3,5-Dimethylbenzenethiol
EINECSNumber (3)-1094
TSCA Yes