missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethylbenzenesulfonyl chloride, 97%

Catalog No. p-7047305
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1860303 1 g
AAL1860306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1860303 Supplier Thermo Scientific Chemicals Supplier No. L1860303

CAS: 2905-27-3 | C8H9ClO2S | 204.67 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2905-27-3
Molecular Formula C8H9ClO2S
Molecular Weight (g/mol) 204.67
MDL Number MFCD03094655
InChI Key LSAGRAXLOLZVKO-UHFFFAOYSA-N
Synonym 3,5-dimethylbenzene-1-sulfonyl chloride, 3,5-dimethylphenyl chlorosulfone, 3,5-dimethylbenzenesulfonylchloride, 3,5-dimethylbenzenesulphonyl chloride, 3,5-dimethyl-benzenesulfonyl chloride, pubchem5129, acmc-1ccrc, 5-chlorosulphonyl-m-xylene, 3,5-dimethylphenylsulfonyl chloride, 3,5-xylenesulfonyl chloride
PubChem CID 2736235
SMILES CC1=CC(=CC(C)=C1)S(Cl)(=O)=O

Specifications

Melting Point 92°C to 94°C
Quantity 1 g
UN Number UN3261
Beilstein 2640617
IUPAC Name 3,5-dimethylbenzenesulfonyl chloride
Formula Weight 204.68
Percent Purity 97%
Chemical Name or Material 3,5-Dimethylbenzenesulfonyl chloride
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only