missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethylaniline 98.0+%, TCI America™

Catalog No. p-7126362
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D067125ML 25 mL
D0671500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D067125ML Supplier TCI America Supplier No. D067125ML
Only null left

Chemical Identifiers

CAS 108-69-0
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.183
MDL Number MFCD00007813
InChI Key MKARNSWMMBGSHX-UHFFFAOYSA-N
Synonym 3,5-xylidine, benzenamine, 3,5-dimethyl, 3,5-xylylamine, 3,5-dimethylphenylamine, 3,5-xylidene, 5-amino-1,3-xylene, 1-amino-3,5-dimethylbenzene, 3,5-dimethylbenzenamine, 3,5-dimethylbenzeneamine, 5-amino-1,3-dimethylbenzene
PubChem CID 7949
IUPAC Name 3,5-dimethylaniline
SMILES CC1=CC(=CC(=C1)N)C

Specifications

Color Yellow
Quantity 25 mL
Physical Form Clear Liquid at 20°C
UN Number 1711
Boiling Point 221°C
Formula Weight 121.18
Percent Purity ≥98.0% (GC,T)
Chemical Name or Material 3,5-Dimethylaniline
EINECSNumber (3)-0129
RTECSNumber ZE9625000
TSCA Yes