missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,5-Dimethylaniline 98.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Quantity | 500 mL |
| Physical Form | Clear Liquid at 20°C |
| UN Number | 1711 |
| Boiling Point | 221°C |
| Formula Weight | 121.18 |
| Percent Purity | ≥98.0% (GC,T) |
| Grade | Not Available |
| Chemical Name or Material | 3,5-Dimethylaniline |
Safety and Handling
| EINECSNumber | (3)-0129 |
| RTECSNumber | ZE9625000 |
| TSCA | Yes |