missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethylaniline, 97+%

Catalog No. AC160195000
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC160195000 500 g Glass bottle
AC160191000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC160195000 Supplier Thermo Scientific Chemicals Supplier No. 160195000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 108-69-0 | C8H11N | 121.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 108-69-0
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.18
MDL Number MFCD00007813
InChI Key MKARNSWMMBGSHX-UHFFFAOYSA-N
Synonym 3,5-xylidine, benzenamine, 3,5-dimethyl, 3,5-xylylamine, 3,5-dimethylphenylamine, 3,5-xylidene, 5-amino-1,3-xylene, 1-amino-3,5-dimethylbenzene, 3,5-dimethylbenzenamine, 3,5-dimethylbenzeneamine, 5-amino-1,3-dimethylbenzene
PubChem CID 7949
IUPAC Name 3,5-dimethylaniline
SMILES CC1=CC(=CC(=C1)N)C

Specifications

Melting Point 9.8°C
CAS Min % 97.5
Color Brown to Brown-Yellow
CAS Max % 100.0
Density 0.9700g/mL
Quantity 500 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Liquid
Refractive Index 1.5560 to 1.558
Assay Percent Range 97% min. (GC)
Specific Gravity 0.97
Linear Formula (CH3)2C6H3NH2
Beilstein 12, 1131
Merck Index 15, 10282
Boiling Point 221°C
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alcohol,ether,carbon tetrachloride
Formula Weight 121.18
Percent Purity 97+%
Chemical Name or Material 3, 5-Dimethylaniline, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a po
EINECSNumber 203-607-0
RTECSNumber ZE9625000
TSCA TSCA
Recommended Storage product darkens if exposed to light and air

RUO – Research Use Only