missing translation for 'onlineSavingsMsg'
Learn More

3',5'-Dimethylacetophenone 97.0+%, TCI America™

Catalog No. D48681G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D48681G 1 g
D48685G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D48681G Supplier TCI America Supplier No. D48681G

Chemical Identifiers

CAS 5379-16-8
Molecular Formula C10H12O
Molecular Weight (g/mol) 148.205
MDL Number MFCD01075693
InChI Key BKIHFZLJJUNKMZ-UHFFFAOYSA-N
Synonym 1-3,5-dimethylphenyl ethanone, 3,5-dimethylacetophenone, 3',5'-dimethylacetophenone, 1-3,5-dimethylphenyl ethan-1-one, ethanone, 1-dimethylphenyl, ethanone, 1-3,5-dimethylphenyl, 1-dimethylphenyl ethan-1-one, pubchem24402, 3', 5'-dimethylacetophenone, 3',5'-dimethyl acetophenone
PubChem CID 14921
IUPAC Name 1-(3,5-dimethylphenyl)ethanone
SMILES CC1=CC(=CC(=C1)C(=O)C)C

Specifications

Color White
Boiling Point 237°C
Quantity 1 g
Formula Weight 148.21
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 3′,5′-Dimethylacetophenone