missing translation for 'onlineSavingsMsg'
Learn More

3',5'-Dimethylacetophenone, 97%

Catalog No. p-7046634
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1548003 1 g
AAL1548006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1548003 Supplier Thermo Scientific Chemicals Supplier No. L1548003

CAS: 5379-16-8 | C10H12O | 148.205 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5379-16-8
Molecular Formula C10H12O
Molecular Weight (g/mol) 148.205
MDL Number MFCD01075693
InChI Key BKIHFZLJJUNKMZ-UHFFFAOYSA-N
Synonym 1-3,5-dimethylphenyl ethanone, 3,5-dimethylacetophenone, 3',5'-dimethylacetophenone, 1-3,5-dimethylphenyl ethan-1-one, ethanone, 1-dimethylphenyl, ethanone, 1-3,5-dimethylphenyl, 1-dimethylphenyl ethan-1-one, pubchem24402, 3', 5'-dimethylacetophenone, 3',5'-dimethyl acetophenone
PubChem CID 14921
IUPAC Name 1-(3,5-dimethylphenyl)ethanone
SMILES CC1=CC(=CC(=C1)C(=O)C)C

Specifications

Melting Point ∼20°C
Density 0.988
Boiling Point 238°C
Flash Point 93°C (199°F)
Odor Characteristic
Refractive Index 1.528
Quantity 1 g
Beilstein 2240987
Formula Weight 148.21
Percent Purity 97%
Chemical Name or Material 3',5'-Dimethylacetophenone
Show More Show Less
Hazard Category H227
Precautionary Statement P210-P280-P370+P378q-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only