missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethyl-p-anisic acid, 98%, Thermo Scientific™

Catalog No. p-115535
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC187130050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC187130050 Supplier Thermo Scientific Chemicals Supplier No. 187130050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 21553-46-8
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.2
MDL Number MFCD00020309
InChI Key WXVQURJGDUNJCS-UHFFFAOYSA-N
Synonym 3,5-dimethyl-4-methoxybenzoic acid, 3,5-dimethyl-p-anisic acid, 4-methoxy-3,5-dimethyl-benzoic acid, 3,5-dimethyl anisic acid, benzoic acid, 4-methoxy-3,5-dimethyl, 4-methoxy-3,5-dimethylbenzoicacid, acmc-209flo, ksc496g8f, 3,5-dimethyl-4-methoxy-benzoic acid, 4-methoxy-3,5-dimethyl benzoic acid
PubChem CID 88944
IUPAC Name 4-methoxy-3,5-dimethylbenzoic acid
SMILES CC1=CC(=CC(=C1OC)C)C(=O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 190°C to 194°C
Color White to Brown
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula CH3OC6H2(CH3)2CO2H
Quantity 5 g
Formula Weight 180.2
Percent Purity 98%
Physical Form Crystalline Solid
Chemical Name or Material 3, 5-Dimethyl-p-anisic acid, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective gloves/protective clothing/eye protection/face protection.
EINECSNumber 244-441-9
TSCA TSCA

RUO – Research Use Only