missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethyl-4-propoxyphenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. p-7193813
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D50801G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D50801G Supplier TCI America Supplier No. D50801G
Only null left

Specifications

Chemical Name or Material 3,5-Dimethyl-4-propoxyphenylboronic Acid (contains varying amounts of Anhydride)
Melting Point 187°C
CAS 357611-51-9
Color White
Quantity 1 g
Physical Form Crystal-Powder at 20°C
Molecular Formula C11H17BO3
MDL Number MFCD05865178
Synonym 3,5-dimethyl-4-propoxyphenyl boronic acid, 4-propoxy-3,5-dimethylphenylboronic acid, boronicacid, b-3,5-dimethyl-4-propoxyphenyl, 3,5-dimethyl-4-n-propoxybenzeneboronic acid, 4-propoxy-3,5-dimethylbenzeneboronic acid, acmc-209ij1, 3,5-dimethyl-4-propoxyphenylboronicacid, 3,5-dimethyl-4-propoxybenzeneboronic acid, 3,5-dimethyl-4-propoxy-phenyl boronic acid, contains varying amounts of anhydride
InChI Key ISUPZUFVQLUFLM-UHFFFAOYSA-N
SMILES CCCOC1=C(C)C=C(C=C1C)B(O)O
IUPAC Name (3,5-dimethyl-4-propoxyphenyl)boronic acid
Molecular Weight (g/mol) 208.06
PubChem CID 16217512
Formula Weight 208.06
Show More Show Less