missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethyl-4-nitrobenzoic acid, 97%

Catalog No. AAL0270403
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0270403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0270403 Supplier Thermo Scientific Chemicals Supplier No. L0270403
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3095-38-3 | C9H9NO4 | 195.17 g/mol

3,5-Dimethyl-4-nitrobenzoic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Dimethyl-4-nitrobenzoic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed in a cool, dry and well ventilated place.

Chemical Identifiers

CAS 3095-38-3
Molecular Formula C9H9NO4
Molecular Weight (g/mol) 195.17
MDL Number MFCD00014699
InChI Key RBAVFNOGEPCOQI-UHFFFAOYSA-N
PubChem CID 249413
IUPAC Name 3,5-dimethyl-4-nitrobenzoic acid
SMILES CC1=CC(=CC(C)=C1[N+]([O-])=O)C(O)=O

Specifications

Melting Point 222°C to 226°C
Odor Odorless
Quantity 1 g
Beilstein 1965772
Synonym 3,5-dimethyl-4-nitrobenzoic acid, benzoic acid, 3,5-dimethyl-4-nitro, 4-nitro-3,5-dimethylbenzoic acid, 3,5-dimethyl-4-nitro-benzoic acid, 4-nitromesitylenic acid, pubchem19635, acmc-1agmi, benzoic acid,5-dimethyl-4-nitro, benzoic acid,3,5-dimethyl-4-nitro, 3,5-dimethyl-4-nitro benzoic acid
Solubility Information Insoluble in water.
Formula Weight 195.18
Percent Purity 97%
Chemical Name or Material 3,5-Dimethyl-4-nitrobenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only