missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethyl-4-isoxazolecarbaldehyde, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC00604DA

Quantity 1g

 View more versions of this product

Catalog No. CC00604DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

54593-26-9
125.127
3,5-dimethyl-4-isoxazolecarbaldehyde, 3,5-dimethylisoxazole-4-carbaldehyde, 3,5-dimethylisoxazole-4-carboxaldehyde, 4-isoxazolecarboxaldehyde, 3,5-dimethyl, 3,5-dimethyl-4-isoxazolecarboxaldehyde, 3,5-dimethyl-4-formalisoxazole, pubchem8682, acmc-1axd1, ksc496e3j, 3,5-dimethylisoxazol-4-carbaldehyde
3,5-dimethyl-1,2-oxazole-4-carbaldehyde
C6H7NO2
TVAYXKLCEILMEA-UHFFFAOYSA-N
289576
CC1=C(C(=NO1)C)C=O

Specifications

3, 5-Dimethyl-4-isoxazolecarbaldehyde
97%
C6H7NO2
3,5-dimethyl-4-isoxazolecarbaldehyde, 3,5-dimethylisoxazole-4-carbaldehyde, 3,5-dimethylisoxazole-4-carboxaldehyde, 4-isoxazolecarboxaldehyde, 3,5-dimethyl, 3,5-dimethyl-4-isoxazolecarboxaldehyde, 3,5-dimethyl-4-formalisoxazole, pubchem8682, acmc-1axd1, ksc496e3j, 3,5-dimethylisoxazol-4-carbaldehyde
CC1=C(C(=NO1)C)C=O
125.127
125.13
54593-26-9
Amber Glass Bottle
1g
TVAYXKLCEILMEA-UHFFFAOYSA-N
3,5-dimethyl-1,2-oxazole-4-carbaldehyde
289576
97%
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents