missing translation for 'onlineSavingsMsg'
Learn More
Learn More
[[(3,5-Dimethyl-1H-pyrrol-2-yl)(3,5-dimethyl-2H-pyrrol-2-ylidene)methyl]methane](difluoroborane) 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 257°C |
| Quantity | 1 g |
| Formula Weight | 262.11 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | [[(3,5-Dimethyl-1H-pyrrol-2-yl)(3,5-dimethyl-2H-pyrrol-2-ylidene)methyl]methane](difluoroborane) |
Safety and Handling
| TSCA | No |