accessibility menu, dialog, popup

UserName

BNNH3CH3CCH3CH3CH3FF

CAS RN 121207-31-6

IUPAC Name: 2,2-difluoro-4,6,8,10,12-pentamethyl-1λ⁵,3-diaza-2-boratricyclo[7.3.0.0³,⁷]dodeca-1(12),4,6,8,10-pentaen-1-ylium-2-uide
Synonyms: 2,2-difluoro-4,6,8,10,12-pentamethyl-1λ⁵,3-diaza-2-boratricyclo[7.3.0.0³,⁷]dodeca-1(12),4,6,8,10-pentaen-1-ylium-2-uide
Molecular Weight (g/mol): 262.11
Molecular Formula: C14H17BF2N2
InChi Key: DRJHPEGNOPSARR-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C2N1[B-](F)(F)[N+]1=C(C)C=C(C)C1=C2C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results