missing translation for 'onlineSavingsMsg'
Learn More

(3,5-Dimethyl-1-phenyl-1H-pyrazol-4-yl)methanol, ≥97%, Thermo Scientific™

Catalog No. p-3175092
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC48009DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC48009DA Supplier Thermo Scientific Chemicals Supplier No. CC48009DA

Chemical Identifiers

CAS 58789-53-0
Molecular Formula C12H14N2O
Molecular Weight (g/mol) 202.257
MDL Number MFCD06202725
InChI Key JMWWSNPRAOYPSF-UHFFFAOYSA-N
Synonym 3,5-dimethyl-1-phenyl-1h-pyrazol-4-yl methanol, 3,5-dimethyl-1-phenylpyrazol-4-yl methanol, 1h-pyrazole-4-methanol,3,5-dimethyl-1-phenyl, 3,5-dimethyl-1-phenylpyrazol-4-yl methan-1-ol
PubChem CID 2526674
IUPAC Name (3,5-dimethyl-1-phenylpyrazol-4-yl)methanol
SMILES CC1=C(C(=NN1C2=CC=CC=C2)C)CO

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 202.26
Percent Purity 97%
Chemical Name or Material (3, 5-Dimethyl-1-phenyl-1H-pyrazol-4-yl)methanol
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.