CAS RN 58789-53-0
CAS RN 58789-53-0
IUPAC Name:
(3,5-dimethyl-1-phenyl-1H-pyrazol-4-yl)methanol
Synonyms:
(3,5-dimethyl-1-phenylpyrazol-4-yl)methanol
Molecular Weight (g/mol):
202.26
Molecular Formula:
C12H14N2O
InChi Key:
JMWWSNPRAOYPSF-UHFFFAOYSA-N
SMILES:
CC1=NN(C(C)=C1CO)C1=CC=CC=C1
1
–
2
of
2
results
1
(3,5-Dimethyl-1-phenyl-1H-pyrazol-4-yl)methanol, ≥97%, Thermo Scientific™
CAS: 58789-53-0 Molecular Formula: C12H14N2O Molecular Weight (g/mol): 202.257 MDL Number: MFCD06202725 InChI Key: JMWWSNPRAOYPSF-UHFFFAOYSA-N Synonym: 3,5-dimethyl-1-phenyl-1h-pyrazol-4-yl methanol,3,5-dimethyl-1-phenylpyrazol-4-yl methanol,1h-pyrazole-4-methanol,3,5-dimethyl-1-phenyl,3,5-dimethyl-1-phenylpyrazol-4-yl methan-1-ol PubChem CID: 2526674 IUPAC Name: (3,5-dimethyl-1-phenylpyrazol-4-yl)methanol SMILES: CC1=C(C(=NN1C2=CC=CC=C2)C)CO