missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethyl-1-adamantanemethanol 98.0+%, TCI America™

Catalog No. D33671G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D33671G 1 g
D33675G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D33671G Supplier TCI America Supplier No. D33671G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 26919-42-6
Molecular Formula C13H22O
Molecular Weight (g/mol) 194.318
InChI Key RVWLWJAOIBEWAV-UHFFFAOYSA-N
Synonym 3,5-dimethyladamantan-1-yl methanol, 3,5-dimethyl-1-adamantanemethanol, tricyclo 3.3.1.13,7 decane-1-methanol, 3,5-dimethyl, 3,5-dimethyladamantanyl methan-1-ol, acmc-1cfe2, 1r,3r,5s,7r-3,5-dimethyladamantan-1-yl methanol, 3,5-dimethyl-1-adamantyl methanol, 3,5-dimethyladamant-1-yl methanol, 3,5-dimethyl-1-hydroxymethyladamantane, 1-hydroxymethyl-3,5-dimethyladamantane
PubChem CID 4050097
IUPAC Name (3,5-dimethyl-1-adamantyl)methanol
SMILES CC12CC3CC(C1)(CC(C3)(C2)CO)C

Specifications

Melting Point 60°C
Color White
Boiling Point 115°C
Quantity 1 g
Formula Weight 194.32
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dimethyl-1-adamantanemethanol