CAS RN 26919-42-6
IUPAC Name:
(3,5-dimethyladamantan-1-yl)methanol
Synonyms:
(3,5-dimethyladamantan-1-yl)methanol
Molecular Weight (g/mol):
194.32
Molecular Formula:
C13H22O
InChi Key:
RVWLWJAOIBEWAV-UHFFFAOYNA-N
SMILES:
CC12CC3CC(C)(C1)CC(CO)(C3)C2
1
–
1
of
1
results
1
3,5-Dimethyl-1-adamantanemethanol 98.0+%, TCI America™
CAS: 26919-42-6 Molecular Formula: C13H22O Molecular Weight (g/mol): 194.318 InChI Key: RVWLWJAOIBEWAV-UHFFFAOYSA-N Synonym: 3,5-dimethyladamantan-1-yl methanol,3,5-dimethyl-1-adamantanemethanol,tricyclo 3.3.1.13,7 decane-1-methanol, 3,5-dimethyl,3,5-dimethyladamantanyl methan-1-ol,acmc-1cfe2,1r,3r,5s,7r-3,5-dimethyladamantan-1-yl methanol,3,5-dimethyl-1-adamantyl methanol,3,5-dimethyladamant-1-yl methanol,3,5-dimethyl-1-hydroxymethyladamantane,1-hydroxymethyl-3,5-dimethyladamantane PubChem CID: 4050097 IUPAC Name: (3,5-dimethyl-1-adamantyl)methanol SMILES: CC12CC3CC(C1)(CC(C3)(C2)CO)C