missing translation for 'onlineSavingsMsg'
Learn More

3,5,-dimethyl-1-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-1h-pyrazole, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC68239CB

Quantity 250mg
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. CC68239CB


Description

Description

Specifications

Chemical Identifiers

937796-06-0
298.19
SERWCEBXGSSDRB-UHFFFAOYSA-N
24229763
CC1=NN(C(C)=C1)C1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1
C17H23BN2O2
MFCD09879979
3,5-dimethyl-1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl-1h-pyrazole, 4-3,5-dimethylpyrazol-1-yl phenylboronic acid, pinacol ester, 3,5-dimethyl-1-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl pyrazole, 3,5,-dimethyl-1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl-1h-pyrazole, 3,5-dimethyl-1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl pyrazole, 4-3,5-dimethyl-1h-pyrazol-1-yl benzeneboronic acid,pinacol ester, 3,5-dimethyl-1-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl-1h-pyrazole
3,5-dimethyl-1-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]pyrazole

Specifications

3,5,-Dimethyl-1-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-1H-pyrazole
96.5
97%
C17H23BN2O2
MFCD09879979
SERWCEBXGSSDRB-UHFFFAOYSA-N
3,5-dimethyl-1-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]pyrazole
24229763
97%
937796-06-0
100.0
Amber Glass Bottle
250mg
3,5-dimethyl-1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl-1h-pyrazole, 4-3,5-dimethylpyrazol-1-yl phenylboronic acid, pinacol ester, 3,5-dimethyl-1-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl pyrazole, 3,5,-dimethyl-1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl-1h-pyrazole, 3,5-dimethyl-1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl pyrazole, 4-3,5-dimethyl-1h-pyrazol-1-yl benzeneboronic acid,pinacol ester, 3,5-dimethyl-1-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl-1h-pyrazole
CC1=NN(C(C)=C1)C1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1
298.19
298.19
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents

Promotions

Promotions