accessibility menu, dialog, popup

UserName

BOOH3CCH3H3CH3CNNCH3CH3

CAS RN 937796-06-0

IUPAC Name: 3,5-dimethyl-1-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-1H-pyrazole
Synonyms: 3,5-dimethyl-1-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]pyrazole
Molecular Weight (g/mol): 298.19
Molecular Formula: C17H23BN2O2
InChi Key: SERWCEBXGSSDRB-UHFFFAOYSA-N
SMILES: CC1=NN(C(C)=C1)C1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)