missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethoxytoluene 97.0+%, TCI America™

Catalog No. p-7127349
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
D25265G 5 g
D252625G 25 g
2 options

Catalog No. D25265G

Supplier: TCI America D25265G

Only null left

Chemical Identifiers

CAS 4179-19-5
Molecular Formula C9H12O2
Molecular Weight (g/mol) 152.19
MDL Number MFCD00015435
InChI Key RIZBLVRXRWHLFA-UHFFFAOYSA-N
Synonym 3,5-dimethoxytoluene, orcinol dimethyl ether, benzene, 1,3-dimethoxy-5-methyl, 5-methylresorcinol dimethyl ether, 3,5-dimethoxy toluene, toluene, 3,5-dimethoxy, 1,3-dimethoxy-5-methyl-benzene, pubchem17775, acmc-1ak5i
PubChem CID 77844
IUPAC Name 1,3-dimethoxy-5-methylbenzene
SMILES COC1=CC(OC)=CC(C)=C1

Specifications

Color Yellow
Boiling Point 112°C
Quantity 5 g
Formula Weight 152.19
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,5-Dimethoxytoluene