missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethoxyphenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. p-7127770
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
D35131G 1 g
D35135G 5 g
2 options

Catalog No. D35131G

Supplier: TCI America D35131G

Only null left

Chemical Identifiers

CAS 192182-54-0
Molecular Formula C8H11BO4
Molecular Weight (g/mol) 181.982
MDL Number MFCD03095127
InChI Key XUIURRYWQBBCCK-UHFFFAOYSA-N
PubChem CID 4374257
IUPAC Name (3,5-dimethoxyphenyl)boronic acid
SMILES B(C1=CC(=CC(=C1)OC)OC)(O)O

Specifications

Melting Point 202°C
Quantity 1 g
Synonym 3,5-dimethoxybenzeneboronic acid, 3,5-dimethoxyphenyl boronic acid, 3,5-dimethoxyphenylboronicacid, boronic acid, 3,5-dimethoxyphenyl, pubchem1827, acmc-1c82c, ksc174i6h, 3,5-dimethoxy phenyl boronic acid, 3,5-dimethoxyphenylboronic acid, boronic acid, b-3,5-dimethoxyphenyl
Formula Weight 181.98
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dimethoxyphenylboronic Acid (contains varying amounts of Anhydride)