missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethoxyphenylacetic Acid 98.0+%, TCI America™

Catalog No. p-7127578
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D29501G 1 g
D29505G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D29501G Supplier TCI America Supplier No. D29501G

Chemical Identifiers

CAS 4670-10-4
Molecular Formula C10H11O4
Molecular Weight (g/mol) 195.20
MDL Number MFCD00016827
InChI Key FFPAFDDLAGTGPQ-UHFFFAOYSA-M
Synonym 3,5-dimethoxyphenylacetic acid, 2-3,5-dimethoxyphenyl acetic acid, 3,5-dimethoxyphenyl acetic acid, benzeneacetic acid, 3,5-dimethoxy, 3,5-dimethoxyphenylaceticacid, 3,5-dimethoxybenzeneacetic acid, 3,5-dimethoxy-phenylacetic acid, 3,5-dimethoxy phenylacetic acid, 3,5-dimethoxy phenyl acetic acid
PubChem CID 138316
IUPAC Name 2-(3,5-dimethoxyphenyl)acetic acid
SMILES COC1=CC(=CC(=C1)CC(=O)O)OC

Specifications

Melting Point 104°C
Color White-Yellow
Quantity 1 g
Formula Weight 196.20
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dimethoxyphenylacetic Acid