missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethoxyphenylacetic acid, 98%

Catalog No. AAB2567203
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2567203 1 g
AAB2567206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2567203 Supplier Thermo Scientific Chemicals Supplier No. B2567203

CAS: 4670-10-4 | C8H14O | 126.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4670-10-4
MDL Number MFCD00016827

Specifications

Melting Point 102°C to 104°C
Molecular Formula C8H14O
Quantity 1 g
Synonym 2,4,4-trimethylcyclopentanone, cyclopentanone, 2,4,4-trimethyl, acmc-1aq0h, 1.1.3-trimethylcyclopentanon-4, cyclopentanone,2,4,4-trimethyl, 2,4,4-trimethyl-cyclopentan-1-one, 2,4,4-trimethyl cyclopentanone
InChI Key OXTQEWUBDTVSFB-UHFFFAOYNA-N
SMILES CC1CC(CC1=O)(C)C
IUPAC Name 2,4,4-trimethylcyclopentan-1-one
Molecular Weight (g/mol) 126.20
PubChem CID 107324
Formula Weight 196.2
Percent Purity 98%
Chemical Name or Material 3,5-Dimethoxyphenylacetic acid
Show More Show Less
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only