missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethoxybenzyl Alcohol 98.0+%, TCI America™

Catalog No. D25945G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D25945G 5 g
D259425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D25945G Supplier TCI America Supplier No. D25945G

Chemical Identifiers

CAS 705-76-0
Molecular Formula C9H12O3
Molecular Weight (g/mol) 168.19
MDL Number MFCD00004641
InChI Key AUDBREYGQOXIFT-UHFFFAOYSA-N
Synonym 3,5-dimethoxybenzyl alcohol, 3,5-dimethoxyphenyl methanol, 3,5-dimethoxybenzylalcohol, benzenemethanol, 3,5-dimethoxy, 3,5-dimethoxy benzyl alcohol, 3,5-dimethoxyphenyl methan-1-ol, 3,5-dimethoxybenzyl alcohole, pubchem7732, 3,5-dimethoxy-benzylalcohol, 3,5-dimethoxy benzylalcohol
PubChem CID 69718
IUPAC Name (3,5-dimethoxyphenyl)methanol
SMILES COC1=CC(OC)=CC(CO)=C1

Specifications

Melting Point 50°C
Color White
Boiling Point 171°C
Quantity 5 g
Formula Weight 168.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dimethoxybenzyl Alcohol