missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethoxybenzyl alcohol, 99%

Catalog No. p-7028026
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2513806 5 g
AAB2513814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2513806 Supplier Thermo Scientific Chemicals Supplier No. B2513806

CAS: 705-76-0 | C9H12O3 | 168.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 705-76-0
Molecular Formula C9H12O3
Molecular Weight (g/mol) 168.19
MDL Number MFCD00004641
InChI Key AUDBREYGQOXIFT-UHFFFAOYSA-N
Synonym 3,5-dimethoxybenzyl alcohol, 3,5-dimethoxyphenyl methanol, 3,5-dimethoxybenzylalcohol, benzenemethanol, 3,5-dimethoxy, 3,5-dimethoxy benzyl alcohol, 3,5-dimethoxyphenyl methan-1-ol, 3,5-dimethoxybenzyl alcohole, pubchem7732, 3,5-dimethoxy-benzylalcohol, 3,5-dimethoxy benzylalcohol
PubChem CID 69718
IUPAC Name (3,5-dimethoxyphenyl)methanol
SMILES COC1=CC(OC)=CC(CO)=C1

Specifications

Melting Point 46°C to 50°C
Boiling Point 183°C to 186°C (20 mmHg)
Flash Point >110°C (230°F)
Quantity 5 g
Beilstein 1866305
Formula Weight 168.19
Percent Purity 99%
Chemical Name or Material 3,5-Dimethoxybenzyl alcohol
EINECSNumber 211-888-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only