missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethoxybenzonitrile, 98%

Catalog No. AAB2298914
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2298914 25 g
AAB2298906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2298914 Supplier Thermo Scientific Chemicals Supplier No. B2298914
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 19179-31-8 | C9H9NO2 | 163.176 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 19179-31-8
Molecular Formula C9H9NO2
Molecular Weight (g/mol) 163.176
MDL Number MFCD00001804
InChI Key NVTHWSJNXVDIKR-UHFFFAOYSA-N
Synonym benzonitrile, 3,5-dimethoxy, 3,5-dimethoxy benzonitrile, 3,5-dimethoxybenzenecarbonitrile, pubchem3122, benzonitrile,5-dimethoxy, acmc-209z2h, 3,5-dimethoxybenzo nitrile, 3-10-00-01449 beilstein handbook reference, ksc497i3p, 1-cyano-3,5-dimethoxybenzene
PubChem CID 29482
IUPAC Name 3,5-dimethoxybenzonitrile
SMILES COC1=CC(=CC(=C1)C#N)OC

Specifications

Melting Point 87°C to 89°C
Quantity 25 g
UN Number UN3439
Formula Weight 163.18
Percent Purity 98%
Chemical Name or Material 3,5-Dimethoxybenzonitrile
Hazard Category H302+H312+H332-H315-H319
Hazard Statement GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber 242-858-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only