missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethoxybenzoic Acid 98.0+%, TCI America™

Catalog No. p-7126774
Change view
Click to view available options
Quantity:
25 g
500 g
Catalog No. Quantity
D153425G 25 g
D1534500G 500 g
2 options

Catalog No. D153425G

Supplier: TCI America D153425G

Only null left

Chemical Identifiers

CAS 1132-21-4
Molecular Formula C9H10O4
Molecular Weight (g/mol) 182.18
MDL Number MFCD00002502
InChI Key IWPZKOJSYQZABD-UHFFFAOYSA-N
Synonym benzoic acid, 3,5-dimethoxy, unii-38my78eckk, 3,5-dimethoxy benzoic acid, 38my78eckk, pubchem3121, 3,5-dimethoxybenzoicacid, acmc-1c4zl, 3,5-dimethoxy-benzoic acid, ksc176m0p, benzoicacid, 3,5-dimethoxy
PubChem CID 14332
IUPAC Name 3,5-dimethoxybenzoic acid
SMILES COC1=CC(=CC(OC)=C1)C(O)=O

Specifications

Melting Point 183°C
Quantity 25 g
Formula Weight 182.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dimethoxybenzoic Acid