missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethoxybenzoic Acid 98.0+%, TCI America™

Catalog No. D153425G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D153425G 25 g
D1534500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D153425G Supplier TCI America Supplier No. D153425G

Chemical Identifiers

CAS 1132-21-4
Molecular Formula C9H10O4
Molecular Weight (g/mol) 182.18
MDL Number MFCD00002502
InChI Key IWPZKOJSYQZABD-UHFFFAOYSA-N
Synonym benzoic acid, 3,5-dimethoxy, unii-38my78eckk, 3,5-dimethoxy benzoic acid, 38my78eckk, pubchem3121, 3,5-dimethoxybenzoicacid, acmc-1c4zl, 3,5-dimethoxy-benzoic acid, ksc176m0p, benzoicacid, 3,5-dimethoxy
PubChem CID 14332
IUPAC Name 3,5-dimethoxybenzoic acid
SMILES COC1=CC(=CC(OC)=C1)C(O)=O

Specifications

Melting Point 183°C
Quantity 25 g
Formula Weight 182.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dimethoxybenzoic Acid