missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,5-Dimethoxyaniline 98.0+%, TCI America™
Supplier: TCI America D148425G
Specifications
| 3,5-Dimethoxyaniline | |
| 57°C | |
| C8H11NO2 | |
| 25 g | |
| WNRGWPVJGDABME-UHFFFAOYSA-N | |
| 3,5-dimethoxyaniline | |
| 66301 | |
| ≥98.0% (GC,T) |
| 10272-07-8 | |
| 178°C | |
| MFCD00008392 | |
| benzenamine, 3,5-dimethoxy, 3,5-dimethoxybenzeneamine, 5-aminoresorcinol dimethyl ether, 3,5-dimethoxy-phenylamine, 3,5-dimethoxyanaline, 3,5-dimethoxy aniline, 5-aminoresorcinol dimethylether, 3,5-dimethoxybenzenamine, 3,5-dimethoxyphenylamine, pubchem4423 | |
| COC1=CC(OC)=CC(N)=C1 | |
| 153.18 | |
| 153.18 | |
| Crystalline Powder |
Chemical Identifiers
| 10272-07-8 | |
| 153.18 | |
| WNRGWPVJGDABME-UHFFFAOYSA-N | |
| 66301 | |
| COC1=CC(OC)=CC(N)=C1 |
| C8H11NO2 | |
| MFCD00008392 | |
| benzenamine, 3,5-dimethoxy, 3,5-dimethoxybenzeneamine, 5-aminoresorcinol dimethyl ether, 3,5-dimethoxy-phenylamine, 3,5-dimethoxyanaline, 3,5-dimethoxy aniline, 5-aminoresorcinol dimethylether, 3,5-dimethoxybenzenamine, 3,5-dimethoxyphenylamine, pubchem4423 | |
| 3,5-dimethoxyaniline |
Safety and Handling
EINECSNumber : (3)-0616
TSCA : Yes
Recommended Storage : Refrigerator