missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethoxyaniline 98.0+%, TCI America™

Catalog No. D14845G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D14845G 5 g
D148425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D14845G Supplier TCI America Supplier No. D14845G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 10272-07-8
Molecular Formula C8H11NO2
Molecular Weight (g/mol) 153.18
MDL Number MFCD00008392
InChI Key WNRGWPVJGDABME-UHFFFAOYSA-N
Synonym benzenamine, 3,5-dimethoxy, 3,5-dimethoxybenzeneamine, 5-aminoresorcinol dimethyl ether, 3,5-dimethoxy-phenylamine, 3,5-dimethoxyanaline, 3,5-dimethoxy aniline, 5-aminoresorcinol dimethylether, 3,5-dimethoxybenzenamine, 3,5-dimethoxyphenylamine, pubchem4423
PubChem CID 66301
IUPAC Name 3,5-dimethoxyaniline
SMILES COC1=CC(OC)=CC(N)=C1

Specifications

Melting Point 57°C
Boiling Point 178°C
Quantity 5 g
Formula Weight 153.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dimethoxyaniline
EINECSNumber (3)-0616
TSCA Yes
Recommended Storage Refrigerator