missing translation for 'onlineSavingsMsg'
Learn More

3,5-Diiodosalicylic Acid 98.0+%, TCI America™

Catalog No. p-7126871
Change view
Click to view available options
Quantity:
25 g
250 g
Catalog No. Quantity
D167725G 25 g
D1677250G 250 g
2 options

Catalog No. D167725G

Supplier: TCI America D167725G

Only null left

Chemical Identifiers

CAS 133-91-5
Molecular Formula C7H4I2O3
Molecular Weight (g/mol) 389.915
MDL Number MFCD00002444
InChI Key DHZVWQPHNWDCFS-UHFFFAOYSA-N
Synonym 3,5-diiodosalicylic acid, 3,5-diiodo-2-hydroxybenzoic acid, benzoic acid, 2-hydroxy-3,5-diiodo, 2-hydroxy-3,5-diiodobenzoate, 3,5-diiodosalicylate, 2-hydroxy-3,5-diiodobenzenecarboxylic acid, salicylic acid, 3,5-diiodo, 2-hydroxy-3,5-diiodo-benzoic acid, benzoic acid, 3,5-diiodo-2-hydroxy, 3,5-diiodosalicylicacid
PubChem CID 8631
IUPAC Name 2-hydroxy-3,5-diiodobenzoic acid
SMILES C1=C(C=C(C(=C1I)O)C(=O)O)I

Specifications

Melting Point 230°C
Color White-Yellow
Quantity 25 g
Formula Weight 389.91
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Diiodosalicylic Acid
EINECSNumber (3)-3824
RTECSNumber VO2800000
TSCA Yes