missing translation for 'onlineSavingsMsg'
Learn More

3,5-Diiodosalicylic acid, 97%

Catalog No. p-7024681
Change view
Click to view available options
Quantity:
100 g
500 g
Catalog No. Quantity
AAA1978622 100 g
AAA1978636 500 g
2 options

Catalog No. AAA1978622

Supplier: Thermo Scientific Chemicals A1978622

Only null left

CAS: 133-91-5 | C7H4I2O3 | 389.915 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 133-91-5
Molecular Formula C7H4I2O3
Molecular Weight (g/mol) 389.915
MDL Number MFCD00002444
InChI Key DHZVWQPHNWDCFS-UHFFFAOYSA-N
Synonym 3,5-diiodosalicylic acid, 3,5-diiodo-2-hydroxybenzoic acid, benzoic acid, 2-hydroxy-3,5-diiodo, 2-hydroxy-3,5-diiodobenzoate, 3,5-diiodosalicylate, 2-hydroxy-3,5-diiodobenzenecarboxylic acid, salicylic acid, 3,5-diiodo, 2-hydroxy-3,5-diiodo-benzoic acid, benzoic acid, 3,5-diiodo-2-hydroxy, 3,5-diiodosalicylicacid
PubChem CID 8631
IUPAC Name 2-hydroxy-3,5-diiodobenzoic acid
SMILES C1=C(C=C(C(=C1I)O)C(=O)O)I

Specifications

Melting Point 228°C to 234°C
Quantity 100 g
Beilstein 2615358
Sensitivity Light sensitive
Merck Index 14,3190
Formula Weight 389.91
Percent Purity 97%
Chemical Name or Material 3,5-Diiodosalicylic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 205-124-0
RTECSNumber VO2800000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only